Learn More
Consolidated Plastics 64 Oz - 5 5/8" ID x 6 1/4" H - PPCO Disposable Containers - White, 50/cs
SDP
Catalog No. NC1576690
Encompass
Lightweight disposable containers with tight fitting lids offer excellent chemical resistance and resist freezing or boiling. Low cost for disposing, sturdy enough for reuse. White opaque containers eliminate sight of specimen.
Catalog No. NC1576690 Supplier Consolidated Plastics Supplier No. 042538
Lightweight disposable containers with tight fitting lids offer excellent chemical resistance and resist freezing or boiling. Low cost for disposing, sturdy enough for reuse. White opaque containers eliminate sight of specimen.