Learn More
Organomation 64 Position MULTIVAP Nitrogen Evaporator
SDP
Supplier:  Organomation 11364
Encompass_Preferred
The 64 position MULTIVAP nitrogen blowdown evaporator is a water bath model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which direct the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 8 samples, which allows individual rows to be shut off when evaporating batches of less than 64 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator. The 64 position MULTIVAP parallel evaporator uses a heated water bath to reach evaporative temperatures ranging from 30°C to 100°C. An unheated version is available for evaporations that will be performed at ambient temperatures. Sample racks are customized within a range of 11-30 mm for the user’s sample size.
Catalog No. NC2220098
Add to Cart
Request a Quote