Learn More
Analytical Sales & Services 64 Position MicroCentrifuge Tube Storage Box
Catalog No. 5000806759
Encompass
Advantage™ 64 Position MicroCentrifuge Tube Storage Box \n\n• For 0.5mL - 2mL Tubes\n• Polyurethane Foam Insert \n\nQty/Pkg: 8
Catalog No. 50-008-06759 Supplier Analytical Sales & Services Supplier No. 96S464
Advantage™ 64 Position MicroCentrifuge Tube Storage Box \n\n• For 0.5mL - 2mL Tubes\n• Polyurethane Foam Insert \n\nQty/Pkg: 8