Learn More
Electron Microscopy Sciences ProPlate™ MicroArray Slide 64 Well Silicone Replacement Gasket, 3.5 x 3.5 mm I.D.
SDP
Catalog No. 5034310
Encompass_Preferred
Replacement silicone gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.
I.D.: 3.5 x 3.5 mm
Catalog No. 50-343-10 Supplier Electron Microscopy Sciences Supplier No. 6348458
Replacement silicone gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.
I.D.: 3.5 x 3.5 mm