Learn More
Nelson Jameson Company 3M Petrifilm Aerobic Count Plate, Sample-ready film plate, 1000 plates/case, For total aerobic bacteria enumeration, Fast results in 48 hours, Red indicator dye, Built-in grid
Catalog No. NC2120335
Encompass
3M Petrifilm Aerobic Count Plate, Sample-ready film plate, 1000 plates/case, For total aerobic bacteria enumeration, Fast results in 48 hours, Red indicator dye, Built-in grid
This item is not returnable. View return policy
Catalog No. NC2120335 Supplier Nelson Jameson Company Supplier No. 4603406CY
3M Petrifilm Aerobic Count Plate, Sample-ready film plate, 1000 plates/case, For total aerobic bacteria enumeration, Fast results in 48 hours, Red indicator dye, Built-in grid