Learn More
NEOGEN Petrifilm High-Sensitivity Coliform Count Plates, 50 plates
SDP
Catalog No. NC2874649
Encompass_Preferred
The Neogen Petrifilm Coliform Count Plate provides confirmed results in just 24 hours. By eliminating subsequent confirmation steps required with most traditional reference methods, the Petrifilm Coliform Count Plates can help increase productivity and reduce overall lab costs. This plate is a sample-ready-culture medium system which contains Violet Red Bile (VRB) nutrients, a cold water soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration in food and beverage samples.
Catalog No. NC2874649 Supplier NEOGEN Supplier No. 700002278
The Neogen Petrifilm Coliform Count Plate provides confirmed results in just 24 hours. By eliminating subsequent confirmation steps required with most traditional reference methods, the Petrifilm Coliform Count Plates can help increase productivity and reduce overall lab costs. This plate is a sample-ready-culture medium system which contains Violet Red Bile (VRB) nutrients, a cold water soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration in food and beverage samples.