Learn More
NEOGEN Petrifilm E. coli /Coliform Count Plates, 500 plates
SDP
Catalog No. NC2874648
Encompass_Preferred
The Neogen Petrifilm E. coli /Coliform Count Plate is a sample-ready-culture medium system which contains modified Violet Red Bile (VRB) nutrients, a cold-water-soluble gelling agent, an indicator of glucuronidase activity, 5-bromo-4-chloro-3-indolyl-D-glucuronide (BCIG), and a tetrazolium indicator that facilitates colony enumeration. Neogen Petrifilm E. coli /Coliform Count Plates are used for the enumeration of Escherichia coli (E. coli ) and coliforms in the food and beverage industries. Coliforms are not defined by taxonomy but rather by their biochemical characteristics.
Catalog No. NC2874648 Supplier NEOGEN Supplier No. 700002277
The Neogen Petrifilm E. coli /Coliform Count Plate is a sample-ready-culture medium system which contains modified Violet Red Bile (VRB) nutrients, a cold-water-soluble gelling agent, an indicator of glucuronidase activity, 5-bromo-4-chloro-3-indolyl-D-glucuronide (BCIG), and a tetrazolium indicator that facilitates colony enumeration. Neogen Petrifilm E. coli /Coliform Count Plates are used for the enumeration of Escherichia coli (E. coli ) and coliforms in the food and beverage industries. Coliforms are not defined by taxonomy but rather by their biochemical characteristics.