Learn More
NEOGEN Petrifilm Enterobacteriaceae Count Plates, 50 plates
SDP
Catalog No. NC2874647
Encompass_Preferred
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petrifilm Enterobacteriaceae Count Plate provides results in just 24 hours, enabling you to quickly determine potential sources of contamination. Enterobacteriaceae includes all coliforms as well as potential pathogens such as Salmonella , Shigella and Yersinia .
Catalog No. NC2874647 Supplier NEOGEN Supplier No. 700002275
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petrifilm Enterobacteriaceae Count Plate provides results in just 24 hours, enabling you to quickly determine potential sources of contamination. Enterobacteriaceae includes all coliforms as well as potential pathogens such as Salmonella , Shigella and Yersinia .