Learn More
NEOGEN Petrifilm Enterobacteriaceae Count Plates, 1000 plates
SDP
Catalog No. NC2874646
Encompass_Preferred
The Neogen Petrifilm Enterobacteriaceae Count Plate provides results in just 24 hours, enabling you to quickly determine potential sources of contamination. Enterobacteriaceae includes all coliforms as well as potential pathogens such as Salmonella , Shigella and Yersinia .
Catalog No. NC2874646 Supplier NEOGEN Supplier No. 700002273
The Neogen Petrifilm Enterobacteriaceae Count Plate provides results in just 24 hours, enabling you to quickly determine potential sources of contamination. Enterobacteriaceae includes all coliforms as well as potential pathogens such as Salmonella , Shigella and Yersinia .