Learn More
NEOGEN Petrifilm Aqua Heterotrophic Count Plates, 100 plates
SDP
Catalog No. NC2874610
Encompass_Preferred
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petri?lm Aqua Heterotrophic Count Plates are used for the enumeration of heterotrophic bacteria in bottled water industries. The Petrifilm Aqua Heterotrophic Count Plate is a sample-ready culture medium system, which contains Standard Methods nutrients, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration.
Catalog No. NC2874610 Supplier NEOGEN Supplier No. 700002133
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petri?lm Aqua Heterotrophic Count Plates are used for the enumeration of heterotrophic bacteria in bottled water industries. The Petrifilm Aqua Heterotrophic Count Plate is a sample-ready culture medium system, which contains Standard Methods nutrients, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration.