Learn More
NEOGEN Petrifilm Aqua Heterotrophic Count Plates, 1000 plates
SDP
Supplier:  NEOGEN 700002135
Encompass_Preferred
The Neogen Petrifilm Aqua Heterotrophic Count Plates are used for the enumeration of heterotrophic bacteria in bottled water industries. The Petrifilm Aqua Heterotrophic Count Plate is a sample-ready culture medium system, which contains Standard Methods nutrients, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration.
Catalog No. NC2874612