Learn More
Nelson Jameson Company Lactic Acid Bacteria Count Petrifilm
Catalog No. NC1864575
Encompass
3M™ advanced technology has developed Lactic Acid Bacteria Count Petrifilm that no longer needs the conventional methods gas packs, chamber or CO2 incubators. The self contained anaerobic environment is enabled by oxygen-scavenging technology and oxygen-barrier films. Results in 48 hours of incubation. The proven reliability versus MRS and APT agar methods, but is an all-in-one method for differentiation of gas-producing colonies without Durham tubes or extra steps. It can be used to test a broad range of foods and environmental samples.
Catalog No. NC1864575 Supplier Nelson Jameson Company Supplier No. 4603461 C
3M™ advanced technology has developed Lactic Acid Bacteria Count Petrifilm that no longer needs the conventional methods gas packs, chamber or CO2 incubators. The self contained anaerobic environment is enabled by oxygen-scavenging technology and oxygen-barrier films. Results in 48 hours of incubation. The proven reliability versus MRS and APT agar methods, but is an all-in-one method for differentiation of gas-producing colonies without Durham tubes or extra steps. It can be used to test a broad range of foods and environmental samples.