Learn More
NEOGEN Petrifilm Aqua Heterotrophic Count Plates, 50 plates
SDP
Catalog No. NC2874639
Encompass_Preferred
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petrifilm Lactic Acid Bacteria Count (LAB) Plate is a self-contained, sample-ready-culture-medium system which contains nutrients, selective agents, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration. The Petrifilm LAB Plate contains oxygen scavenging compounds which create an anaerobic environment for the recovery of homofermentative and heterofermentative lactic acid bacteria in the food and beverage industries. .
Catalog No. NC2874639 Supplier NEOGEN Supplier No. 700002228
This item has a minimum order purchase requirement of 2 per supplier requirements
The Neogen Petrifilm Lactic Acid Bacteria Count (LAB) Plate is a self-contained, sample-ready-culture-medium system which contains nutrients, selective agents, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration. The Petrifilm LAB Plate contains oxygen scavenging compounds which create an anaerobic environment for the recovery of homofermentative and heterofermentative lactic acid bacteria in the food and beverage industries. .