Learn More
NEOGEN Petrifilm Aqua Heterotrophic Count Plates, 500 plates
SDP
Supplier:  NEOGEN 700002227
Encompass_Preferred
The Neogen Petrifilm Lactic Acid Bacteria Count (LAB) Plate is a self-contained, sample-ready-culture-medium system which contains nutrients, selective agents, a cold-water-soluble gelling agent, and a tetrazolium indicator that facilitates colony enumeration. The Petrifilm LAB Plate contains oxygen scavenging compounds which create an anaerobic environment for the recovery of homofermentative and heterofermentative lactic acid bacteria in the food and beverage industries. .
Catalog No. NC2874637