Learn More
ABclonal Technology 647 anti-h/m ITGA5/CD49e
SDP
Catalog No. 5000005883
Encompass_Preferred
The product of this gene belongs to the integrin alpha chain family Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha 5 subunit This subunit associates with the beta 1 subunit to form a fibronectin receptor This integrin may promote tumor invasion and higher expression of this gene may be correlated with shorter survival time in lung cancer patients Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes
Catalog No. 50-000-05883 Supplier ABclonal Technology Supplier No. A22772200T
The product of this gene belongs to the integrin alpha chain family Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha 5 subunit This subunit associates with the beta 1 subunit to form a fibronectin receptor This integrin may promote tumor invasion and higher expression of this gene may be correlated with shorter survival time in lung cancer patients Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes