Learn More
ABclonal Technology 647 anti-Human CD119/IFNGR1
SDP
Catalog No. 5000005829
Encompass_Preferred
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2 A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection In addition defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease also known as familial disseminated atypical mycobacterial infection
Catalog No. 50-000-05829 Supplier ABclonal Technology Supplier No. A23817200T
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2 A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection In addition defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease also known as familial disseminated atypical mycobacterial infection