Learn More
ABclonal Technology 647 anti-Human CD90/Thy1
SDP
Catalog No. 5000007114
Encompass_Preferred
This gene encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins The encoded protein is involved in cell adhesion and cell communication in numerous cell types but particularly in cells of the immune and nervous systems The encoded protein is widely used as a marker for hematopoietic stem cells This gene may function as a tumor suppressor in nasopharyngeal carcinoma Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07114 Supplier ABclonal Technology Supplier No. A22511200T
This gene encodes a cell surface glycoprotein and member of the immunoglobulin superfamily of proteins The encoded protein is involved in cell adhesion and cell communication in numerous cell types but particularly in cells of the immune and nervous systems The encoded protein is widely used as a marker for hematopoietic stem cells This gene may function as a tumor suppressor in nasopharyngeal carcinoma Alternative splicing results in multiple transcript variants