Learn More
ABclonal Technology 647 Rabbit anti-Human CD200R
SDP
Supplier:  ABclonal Technology A25711200T
Encompass_Preferred
This gene encodes a receptor for the OX-2 membrane glycoprotein Both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains This receptor is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal Mouse studies of a related gene suggest that this interaction may control myeloid function in a tissue-specific manner Alternative splicing of this gene results in multiple transcript variants
Catalog No. 50-000-12475