Learn More
ABclonal Technology 647 Rabbit anti-Human CD200R
SDP
Catalog No. 5000012474
Encompass_Preferred
This gene encodes a receptor for the OX-2 membrane glycoprotein Both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains This receptor is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal Mouse studies of a related gene suggest that this interaction may control myeloid function in a tissue-specific manner Alternative splicing of this gene results in multiple transcript variants
Catalog No. 50-000-12474 Supplier ABclonal Technology Supplier No. A25711100T
This gene encodes a receptor for the OX-2 membrane glycoprotein Both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains This receptor is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal Mouse studies of a related gene suggest that this interaction may control myeloid function in a tissue-specific manner Alternative splicing of this gene results in multiple transcript variants