Learn More
ABclonal Technology 647 Rabbit anti-Human CD34
SDP
Catalog No. 5000012668
Encompass_Preferred
The protein encoded by this gene may play a role in the attachment of stem cells to the bone marrow extracellular matrix or to stromal cells This single-pass membrane protein is highly glycosylated and phosphorylated by protein kinase C Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12668 Supplier ABclonal Technology Supplier No. A25894100T
The protein encoded by this gene may play a role in the attachment of stem cells to the bone marrow extracellular matrix or to stromal cells This single-pass membrane protein is highly glycosylated and phosphorylated by protein kinase C Two transcript variants encoding different isoforms have been found for this gene