Learn More
ABclonal Technology 647 Rabbit anti-Human CD38
SDP
Catalog No. 5000009754
Encompass_Preferred
The protein encoded by this gene is a non-lineage-restricted type II transmembrane glycoprotein that synthesizes and hydrolyzes cyclic adenosine 5 -diphosphate-ribose an intracellular calcium ion mobilizing messenger The release of soluble protein and the ability of membrane-bound protein to become internalized indicate both extracellular and intracellular functions for the protein This protein has an N-terminal cytoplasmic tail a single membrane-spanning domain and a C-terminal extracellular region with four N-glycosylation sites Crystal structure analysis demonstrates that the functional molecule is a dimer with the central portion containing the catalytic site It is used as a prognostic marker for patients with chronic lymphocytic leukemia Alternative splicing results in multiple transcript variants
Catalog No. 50-000-09754 Supplier ABclonal Technology Supplier No. A24365100T
The protein encoded by this gene is a non-lineage-restricted type II transmembrane glycoprotein that synthesizes and hydrolyzes cyclic adenosine 5 -diphosphate-ribose an intracellular calcium ion mobilizing messenger The release of soluble protein and the ability of membrane-bound protein to become internalized indicate both extracellular and intracellular functions for the protein This protein has an N-terminal cytoplasmic tail a single membrane-spanning domain and a C-terminal extracellular region with four N-glycosylation sites Crystal structure analysis demonstrates that the functional molecule is a dimer with the central portion containing the catalytic site It is used as a prognostic marker for patients with chronic lymphocytic leukemia Alternative splicing results in multiple transcript variants