Learn More
ABclonal Technology 647 Rabbit anti-Human CD45
SDP
Supplier:  ABclonal Technology A22495200T
Encompass_Preferred
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth differentiation mitosis and oncogenic transformation This PTP contains an extracellular domain a single transmembrane segment and two tandem intracytoplasmic catalytic domains and thus is classified as a receptor type PTP This PTP has been shown to be an essential regulator of T- and B-cell antigen receptor signaling It functions through either direct interaction with components of the antigen receptor complexes or by activating various Src family kinases required for the antigen receptor signaling This PTP also suppresses JAK kinases and thus functions as a regulator of cytokine receptor signaling Alternatively spliced transcripts variants of this gene which encode distinct isoforms have been reported
Catalog No. 50-000-07727