Learn More
ABclonal Technology 647 Rabbit anti-Human CD46
SDP
Catalog No. 5000011745
Encompass_Preferred
The protein encoded by this gene is a type I membrane protein and is a regulatory part of the complement system The encoded protein has cofactor activity for inactivation of complement components C3b and C4b by serum factor I which protects the host cell from damage by complement In addition the encoded protein can act as a receptor for the Edmonston strain of measles virus human herpesvirus-6 and type IV pili of pathogenic Neisseria Finally the protein encoded by this gene may be involved in the fusion of the spermatozoa with the oocyte during fertilization Mutations at this locus have been associated with susceptibility to hemolytic uremic syndrome Alternatively spliced transcript variants encoding different isoforms have been described
Catalog No. 50-000-11745 Supplier ABclonal Technology Supplier No. A25551500T
The protein encoded by this gene is a type I membrane protein and is a regulatory part of the complement system The encoded protein has cofactor activity for inactivation of complement components C3b and C4b by serum factor I which protects the host cell from damage by complement In addition the encoded protein can act as a receptor for the Edmonston strain of measles virus human herpesvirus-6 and type IV pili of pathogenic Neisseria Finally the protein encoded by this gene may be involved in the fusion of the spermatozoa with the oocyte during fertilization Mutations at this locus have been associated with susceptibility to hemolytic uremic syndrome Alternatively spliced transcript variants encoding different isoforms have been described