Learn More
ABclonal Technology 647 Rabbit anti-Human CD58
SDP
Catalog No. 5000006814
Encompass_Preferred
This gene encodes a member of the immunoglobulin superfamily The encoded protein is a ligand of the T lymphocyte CD2 protein and functions in adhesion and activation of T lymphocytes The protein is localized to the plasma membrane Alternatively spliced transcript variants have been described
Catalog No. 50-000-06814 Supplier ABclonal Technology Supplier No. A22513200T
This gene encodes a member of the immunoglobulin superfamily The encoded protein is a ligand of the T lymphocyte CD2 protein and functions in adhesion and activation of T lymphocytes The protein is localized to the plasma membrane Alternatively spliced transcript variants have been described