Learn More
ABclonal Technology 647 Rabbit anti-Human CD66b
SDP
Catalog No. 5000006996
Encompass_Preferred
Enables protein heterodimerization activity Involved in heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules Located in cell surface and extracellular space Biomarker of severe acute respiratory syndrome
Catalog No. 50-000-06996 Supplier ABclonal Technology Supplier No. A22686200T
Enables protein heterodimerization activity Involved in heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules Located in cell surface and extracellular space Biomarker of severe acute respiratory syndrome