Learn More
ABclonal Technology 647 Rabbit anti-Human CD9
SDP
Catalog No. 5000005496
Encompass_Preferred
This gene encodes a member of the transmembrane 4 superfamily also known as the tetraspanin family Tetraspanins are cell surface glycoproteins with four transmembrane domains that form multimeric complexes with other cell surface proteins The encoded protein functions in many cellular processes including differentiation adhesion and signal transduction and expression of this gene plays a critical role in the suppression of cancer cell motility and metastasis
Catalog No. 50-000-05496 Supplier ABclonal Technology Supplier No. A24157200T
This gene encodes a member of the transmembrane 4 superfamily also known as the tetraspanin family Tetraspanins are cell surface glycoproteins with four transmembrane domains that form multimeric complexes with other cell surface proteins The encoded protein functions in many cellular processes including differentiation adhesion and signal transduction and expression of this gene plays a critical role in the suppression of cancer cell motility and metastasis