Learn More
ABclonal Technology 647 Rabbit anti-Human CD98
Supplier: ABclonal Technology A24933200T
This gene is a member of the solute carrier family and encodes a cell surface transmembrane protein The protein exists as the heavy chain of a heterodimer covalently bound through di-sulfide bonds to one of several possible light chains The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids Alternatively spliced transcript variants encoding different isoforms have been characterized