Learn More
ABclonal Technology 647 Rabbit anti-Human FATP1
SDP
Catalog No. 5000012327
Encompass_Preferred
Enables biotin transmembrane transporter activity efflux transmembrane transporter activity and long-chain fatty acid transporter activity Involved in several processes including carboxylic acid transmembrane transport glycerophospholipid biosynthetic process and lipid transport across blood-brain barrier Located in membrane Part of plasma membrane
Catalog No. 50-000-12327 Supplier ABclonal Technology Supplier No. A25571100T
Enables biotin transmembrane transporter activity efflux transmembrane transporter activity and long-chain fatty acid transporter activity Involved in several processes including carboxylic acid transmembrane transport glycerophospholipid biosynthetic process and lipid transport across blood-brain barrier Located in membrane Part of plasma membrane