Learn More
ABclonal Technology 647 Rabbit anti-Human GATA3
SDP
Supplier:  ABclonal Technology A25946100T
Encompass_Preferred
This gene encodes a protein which belongs to the GATA family of transcription factors The protein contains two GATA-type zinc fingers and is an important regulator of T-cell development and plays an important role in endothelial cell biology Defects in this gene are the cause of hypoparathyroidism with sensorineural deafness and renal dysplasia
Catalog No. 50-000-13162