Learn More
ABclonal Technology 647 Rabbit anti-Human GP6/GPVI
SDP
Catalog No. 5000005605
Encompass_Preferred
This gene encodes a platelet membrane glycoprotein of the immunoglobulin superfamily The encoded protein is a receptor for collagen and plays a critical role in collagen-induced platelet aggregation and thrombus formation The encoded protein forms a complex with the Fc receptor gamma-chain that initiates the platelet activation signaling cascade upon collagen binding Mutations in this gene are a cause of platelet-type bleeding disorder-11 (BDPLT11) Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene
Catalog No. 50-000-05605 Supplier ABclonal Technology Supplier No. A24104100T
This gene encodes a platelet membrane glycoprotein of the immunoglobulin superfamily The encoded protein is a receptor for collagen and plays a critical role in collagen-induced platelet aggregation and thrombus formation The encoded protein forms a complex with the Fc receptor gamma-chain that initiates the platelet activation signaling cascade upon collagen binding Mutations in this gene are a cause of platelet-type bleeding disorder-11 (BDPLT11) Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene