Learn More
ABclonal Technology 647 Rabbit anti-Human IgE
SDP
Supplier:  ABclonal Technology A23697100T
Encompass_Preferred
IgE is the class of antibodies produced in the lungs skin and mucous membranes It may protect against parasite invasion but it is a major factor in allergic reactions The antigen-specific IgE interacts with mast cells and eosinophils triggers the release of histamine leukotrienes and other substances that lead to the itching sneezing and congestion of allergies - and the life threatening respiratory distress of asthma and anaphylactic shock
Catalog No. 50-000-06054