Learn More
ABclonal Technology 647 Rabbit anti-Human MERTK
Supplier: ABclonal Technology A24728100T
This gene is a member of the MER AXL TYRO3 receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains two Ig-like C2-type (immunoglobulin-like) domains and one tyrosine kinase domain Mutations in this gene have been associated with disruption of the retinal pigment epithelium (RPE) phagocytosis pathway and onset of autosomal recessive retinitis pigmentosa (RP)