Learn More
ABclonal Technology 647 Rabbit anti-Human NG2
SDP
Catalog No. 5000011971
Encompass_Preferred
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells
Catalog No. 50-000-11971 Supplier ABclonal Technology Supplier No. A24979200T
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells