Learn More
ABclonal Technology 647 Rabbit anti-Human TIGIT
Supplier: ABclonal Technology A23354500T
This gene encodes a member of the PVR (poliovirus receptor) family of immunoglobin proteins The product of this gene is expressed on several classes of T cells including follicular B helper T cells (TFH) The protein has been shown to bind PVR with high affinity this binding is thought to assist interactions between TFH and dendritic cells to regulate T cell dependent B cell responses