Learn More
ABclonal Technology 647 Rabbit anti-Human TROP-2
SDP
Supplier:  ABclonal Technology A22578100T
Encompass_Preferred
This intronless gene encodes a carcinoma-associated antigen This antigen is a cell surface receptor that transduces calcium signals Mutations of this gene have been associated with gelatinous drop-like corneal dystrophy
Catalog No. 50-000-07149