Learn More
Vector Laboratories Horse Anti-Mouse, DyLight 594 DI-2649-1.5
SDP
Supplier:  Vector Laboratories DI26491.5
Encompass
Horse Anti-Mouse IgG Antibody (H+L), DyLight 594, 1.5 mg, is an affinity-purified, ultrapure, high-affinity antibody that has been thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 594 to provide the brightest label for fluorescence microscopy. Supplied in solution. Excitation: 592 nm; Emission: 617 nm; Color: Red.
Catalog No. NC1601052