Learn More
NEOGEN Petrifilm Environmental Listeria Plates, 20 disks
SDP
Supplier:  NEOGEN 700002142
Encompass_Preferred
This item has a minimum order purchase requirement of 6 per supplier requirements
Used when background flora exists in your Staphylococcus testing. This disk is used to identify Staphylococcus aureus from all suspect colonies.
Catalog No. NC2874616