Learn More
Ace Glass, Inc. 10mm 64cm stainless steel stir shaft, PTFE covered, drilled type
SDP
Catalog No. NC0131555
Encompass_Preferred
64cm long PTFE-covered, 10mm O.D. stainless steel drilled-type stirring shaft. Can be used unlubricated with a water-cooled Trubore bearing to achieve 500 rpm maximum. When used with a Trubore bearing lubricated with stir-lube, 1500 rpm maximum can be achieved. When used with a condensate film as an added heat transfer agent, 2000 rpm maximum can be achieved.
Catalog No. NC0131555 Supplier Ace Glass, Inc. Supplier No. 807107
64cm long PTFE-covered, 10mm O.D. stainless steel drilled-type stirring shaft. Can be used unlubricated with a water-cooled Trubore bearing to achieve 500 rpm maximum. When used with a Trubore bearing lubricated with stir-lube, 1500 rpm maximum can be achieved. When used with a condensate film as an added heat transfer agent, 2000 rpm maximum can be achieved.