Learn More
Simco 64IN PLSD/STDY STATE STND
Catalog No. NC3353862
Encompass_Preferred
64" Pulsed & Steady State Standalone AeroBar (24VAC) with FMS Alarm Output and Titanium Emitter Points (13), RoHS. Requires Handheld Remote Below (91-5570) (No Interface Module Required)
Catalog No. NC3353862 Supplier Simco Supplier No. 15225SC64R
64" Pulsed & Steady State Standalone AeroBar (24VAC) with FMS Alarm Output and Titanium Emitter Points (13), RoHS. Requires Handheld Remote Below (91-5570) (No Interface Module Required)