Learn More
Uline Cold Bricks
Catalog No. NC0713677
Encompass
Foam bricks maintain their shape either frozen or thawed, Freeze before use, Recommended for tight packaging situations, Nontoxic heavy-duty plastic pouch 5 mil poly, FDA compliant, Solid when frozen not flexible, May be reused if internal foam core is not broken or cracked, Can be used to prevent freezing if heated in a microwave, Heat on low setting for 30 second increments, Flip pack over every 30 seconds until warm, 100 Recyclable, Outer bag can be recycled, Inside floral foam will break down in a landfill, Do not consume, Do no apply to the body
Catalog No. NC0713677 Supplier Uline Supplier No. S14296
Foam bricks maintain their shape either frozen or thawed, Freeze before use, Recommended for tight packaging situations, Nontoxic heavy-duty plastic pouch 5 mil poly, FDA compliant, Solid when frozen not flexible, May be reused if internal foam core is not broken or cracked, Can be used to prevent freezing if heated in a microwave, Heat on low setting for 30 second increments, Flip pack over every 30 seconds until warm, 100 Recyclable, Outer bag can be recycled, Inside floral foam will break down in a landfill, Do not consume, Do no apply to the body