Learn More
US Plastic Corporation 64 oz. White Fluorinated HDPE F-Style Jug with 38/400 Neck
SDP
Catalog No. NC1919010
Encompass
These F-Style jugs are ideal for packaging challenging liquids, such as chemicals or oil-based solvent products, that would otherwise require metal or glass containers. The in-mold fluorination process enables these lightweight, HDPE jugs to be used in applications involving insecticides, herbicides, cleaning solvents, agricultural chemicals, and petroleum-based products. In-line Fluorinated AIROPAK Barrier Jugs are equal to, if not better than, Post Treated Level 5 containers.
Catalog No. NC1919010 Supplier US Plastic Corporation Supplier No. 68685
These F-Style jugs are ideal for packaging challenging liquids, such as chemicals or oil-based solvent products, that would otherwise require metal or glass containers. The in-mold fluorination process enables these lightweight, HDPE jugs to be used in applications involving insecticides, herbicides, cleaning solvents, agricultural chemicals, and petroleum-based products. In-line Fluorinated AIROPAK Barrier Jugs are equal to, if not better than, Post Treated Level 5 containers.