Learn More
Electron Microscopy Sciences Cooling Chamber 64-Place Step
SDP
Catalog No. 501899060
Encompass_Preferred
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe. Fits 64 1.5 ml microtubes (16 holes per 'step').
Catalog No. 50-189-9060 Supplier Electron Microscopy Sciences Supplier No. 6195530
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe. Fits 64 1.5 ml microtubes (16 holes per 'step').