Learn More
Chemglass Life Sciences 65/40 Ball Joint, Clamp Size: 65, Tubing OD: 46mm
SDP
Catalog No. CG12013
Encompass_Preferred
65/40 Ball Joint, Clamp Size: 65, Tubing OD: 46mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12013 Supplier Chemglass Life Sciences Supplier No. CG12013
65/40 Ball Joint, Clamp Size: 65, Tubing OD: 46mm. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.