Learn More
Chemglass Life Sciences 65/40 O-Ring Ball Joint, Clamp Size: 65, O-Ring Size: 137, Tubing OD: 46mm
SDP
Catalog No. CG12211
Encompass_Preferred
65/40 O-Ring Ball Joint, Clamp Size: 65, O-Ring Size: 137,Tubing OD: 46mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12211 Supplier Chemglass Life Sciences Supplier No. CG12211
65/40 O-Ring Ball Joint, Clamp Size: 65, O-Ring Size: 137,Tubing OD: 46mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.