Learn More
Ace Glass, Inc. 65/40 SOCKET JOINT (GLASSBLOWER)
SDP
Catalog No. 50979414
Encompass
Full length, standard wall 65/40 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-979-414 Supplier Ace Glass, Inc. Supplier No. 765566
Full length, standard wall 65/40 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.