Learn More
Chemglass Life Sciences Joint, 65/40, Socket, Clamp Size 65, 46mm Tubing
SDP
Catalog No. CG12113
Encompass_Preferred
Joint, 65/40, Socket, Clamp Size 65, 46mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12113 Supplier Chemglass Life Sciences Supplier No. CG12113
Joint, 65/40, Socket, Clamp Size 65, 46mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.