Learn More
National Safety Apparel™ 65 CAL ArcGuard™ Arc Flash Hood 14 oz. Brown Kelvar/Nomex
SDP
Catalog No. 17001724 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Caramel
Catalog No.
17-001-724 Color: Caramel
1 options
Catalog No. 17-001-724 Supplier National Safety Apparel™ Supplier No. H65KDQTHHPV
Inherently flame-resistant so they won’t burn, support combustion, or melt
  • Arc Rating: 65 cal/cm²
  • CAT 4
  • Fabric/Material: Multilayer DuPont™ Nomex™ and Kevlar™
  • PureView™ faceshield technology features a greater range of visibility
  • One size fits all with the universal adapter
  • FR fabric will not melt, drip, or ignite
  • Breathable FR fabric
  • Inherently flame resistant for FR protection that will not wash out
  • Hard hat included
  • Premium anti-fog & scratch resistant coating on faceshield
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™, Kevlar™
Color Caramel
Seam Type Stitched
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269