Learn More
National Safety Apparel™ 65 Cal ArcGuard™ Nomex/Kevlar 49 in. Arc Flash Coat
SDP
Catalog No. 17001181 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-181 Small
17-001-182 Medium
17-001-183 Large
17-001-184 2X-Large
17-001-185 3X-Large
17-001-186 X-Large
17-001-187 4X-Large
17-001-188 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-181 Supplier National Safety Apparel™ Supplier No. C04KDQT03SM49
Add to Cart
Add to Cart
Multi-layer Nomex™/Kevlar™ arc flash coat
  • Multi-layer Nomex™ and Kevlar™ material
  • 49 in. Long
  • Stand-up collar
  • FR hook and loop placket closure
  • FR knit wrist
  • No metal closures
  • Generous cut
  • Meets OSHA 1910.269 standards
  • Available in convenient kit (KIT4LC65)
Specifications
Closure Type FR Hook and Loop
Color Brown
Disposable Reusable
Gender Unisex
Material DuPont™ Nomex™, Kevlar™
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269