Learn More
National Safety Apparel™ 65 Cal ArcGuard™ Nomex/Kevlar Arc Flash Coat
SDP
Catalog No. 17001180 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-180 5X-Large
17-001-173 Small
17-001-174 Medium
17-001-175 Large
17-001-176 2X-Large
17-001-177 3X-Large
17-001-178 X-Large
17-001-179 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-180 Supplier National Safety Apparel™ Supplier No. C04KDQT035X32
Kevlar short coat
  • FR Fabric will NOT melt, drip, or ignite
  • FR Hook and loop front closure
  • Inherently Flame Resistant
  • FR Knit wrist cuffs
  • Generous cut allows for unrestrictive movement
Specifications
Closure Type FR Hook and Loop
Color Brown
Disposable Reusable
Gender Unisex
Material Multi-layer DuPont™ Nomex™, Kevlar™
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269