Learn More
National Safety Apparel™ 65 Cal ArcGuard™ Nomex/Kevlar Arc Flash Hood
SDP
Catalog No. 17001723 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Caramel
Catalog No.
17-001-723 Color: Caramel
1 options
Catalog No. 17-001-723 Supplier National Safety Apparel™ Supplier No. H65KDQTFANPV
DuPont™ Nomex™ Brand Fiber Provides lightweight, durable, heat and flame-resistant solutions. Made of 34% aramid, 33% lyocell, 31% modacrylic, and 2% anti-static materials, Nomex™ is engineered to absorb moisture and dry quickly
  • Arc Rating: 65 cal/cm²
  • CAT 4
  • Fabric/Material: Multilayer DuPont™ Nomex™ and Kevlar™
  • Breathable FR fabric Premium anti-fog & scratch resistant coating on faceshield
  • One size fits all with the universal adapter
  • FR fabric will not melt, drip, or ignite
  • Inherently flame resistant, FR protection will not wash out
  • Fans are attached with FR hook and loop
  • Fans are powered by 9-volt batteries
  • PureView™ faceshield technology features a greater range of visibility
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™, Kevlar™
Color Caramel
Seam Type Stitched
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269